missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Aminoguanidine sulfate, 98%
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
Specifications
Specifications
| Quantity | 250 g |
| Formula Weight | 246.24 |
| Percent Purity | 98% |
| Chemical Name or Material | Aminoguanidine sulfate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction