missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine hydrochloride, MedChemExpress

Product Code. 16458576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458576 Supplier Medchem Express Supplier No. HYB1041
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aminoguanidine hydrochloride is a diamine oxidase and NO synthase inhibitor, reduces levels of advanced glycation end products (AGEs) through interacting with 3-deoxyglucosone, is an investigational drug for the treatment of diabetic nephropathy.

Aminoguanidine hydrochloride is a diamine oxidase and NO synthase inhibitor, reduces levels of advanced glycation end products (AGEs) through interacting with 3-deoxyglucosone, is an investigational drug for the treatment of diabetic nephropathy.

Specifications

Chemical Name or Material Aminoguanidine hydrochloride
Physical Form Solid
CAS 1937-19-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula CH7ClN4
Synonym Pimagedine hydrochloride GER-11 Aminoguanidinium chloride
Solubility Information H2O : ≥ 100 mg/mL (904.57 mM) ∣DMSO : 100 mg/mL (904.57 mM; Need ultrasonic)
SMILES NNC(N)=N.[H]Cl
Molecular Weight (g/mol) 110.55
Formula Weight 110.55
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.