missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoguanidine bicarbonate, 98.5%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
1kg
250g
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 125°C |
| Color | White |
| Ignition Residue | 0.3% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% |
| Packaging | Plastic Bottle |
| Linear Formula | H2NNHC(=NH)NH2·H2CO3 |
| Quantity | 1 kg |
| Beilstein | 03,117 |
| Solubility Information | Solubility in water: <5g/L (20°C) |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- Aminoguanidine bicarbonate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Respiratory sensitiser Category 1
- H411-Toxic to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido