missing translation for 'onlineSavingsMsg'
Learn More

(Aminoethylaminomethyl)phenethyltrimethoxysilane, mixture of m and p isomers

Product Code. 11334948
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334948 2 g 2g
11354948 10 g 10g
11344948 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334948 Supplier Thermo Scientific Alfa Aesar Supplier No. 043904.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a crosslinking agent | CAS: 74113-77-2 | H2NC2H4NHCH2C6H4C2H4Si(OCH3)3

As a crosslinking agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74113-77-2
Molecular Formula H2NC2H4NHCH2C6H4C2H4Si(OCH3)3
MDL Number MFCD00054775
Synonym aminoethy-laminomethyl phenethyl-trimethoxysilane, 4-2-aminoethylaminomethyl phenethyltrimethoxysilane, n1-4-2-trimethoxysilyl ethyl benzyl ethane-1,2-diamine, n'-4-2-trimethoxysilylethyl phenyl methyl ethane-1,2-diamine, 2-aminoethyl 4-2-trimethoxysilyl ethyl phenyl methyl amine

Specifications

Boiling Point 128°C (0.2mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 2 g
UN Number UN2735
Flash Point >110°C (230°F)
Formula Weight 298.46
Sensitivity Air and moisture sensitive
Density 1.02 g/mL
Chemical Name or Material (Aminoethylaminomethyl)phenethyltrimethoxysilane, Mixture of m and p isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.