missing translation for 'onlineSavingsMsg'
Learn More

Aminodiphenylmethane, 97%

Product Code. 10551721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10551721 25 g Glass bottle 25g
10676481 1 g Glass bottle 1g
10187893 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10551721 Supplier Thermo Scientific Acros Supplier No. 304320250
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-00-9 | C13H13N | 183.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-00-9
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00008059
InChI Key MGHPNCMVUAKAIE-UHFFFAOYSA-N
Synonym aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine
PubChem CID 7036
IUPAC Name diphenylmethanamine
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)N

Specifications

Melting Point 12.0°C
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 295.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CHNH2
Refractive Index 1.5950 to 1.5970
Quantity 25 g
Beilstein 12, 1323
Merck Index 15, 1078
Specific Gravity 1.06
Formula Weight 183.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Aminodiphenylmethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.