missing translation for 'onlineSavingsMsg'
Learn More

Aminoadipic acid, MedChemExpress

Product Code. 16441199
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16441199

Brand: Medchem Express HY113328

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aminoadipic acid is an intermediate in the metabolism of lysine and saccharopine.

Aminoadipic acid is an intermediate in the metabolism of lysine and saccharopine.

Specifications

Chemical Name or Material Aminoadipic acid
Physical Form Powder
CAS 542-32-5
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C6H11NO4
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information H2O : 5 mg/mL (31.03 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (12.41 mM; Need ultrasonic)
SMILES O=C(O)C(N)CCCC(O)=O
Molecular Weight (g/mol) 161.16
Formula Weight 161.16
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.