missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoacylase/ACY1 Rabbit anti-Human, Clone: 6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89558-100UL
This item is not returnable.
View return policy
Description
Aminoacylase/ACY1 Monoclonal specifically detects Aminoacylase/ACY1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Aminoacylase/ACY1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 95 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunofluorescence | |
| 6 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase | |
| Recombinant Human Aminoacylase/ACY1 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction