missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoacylase/ACY1 Antibody (OTI1D4), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70080PE
This item is not returnable.
View return policy
Description
Aminoacylase/ACY1 Monoclonal specifically detects Aminoacylase/ACY1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Aminoacylase/ACY1 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| ACY1 | |
| Full length recombinant protein of human Aminoacylase/ACY1 (NP_000657) produced in HEK293 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1D4 | |
| Flow Cytometry | |
| ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase | |
| Mouse | |
| Affinity Purified | |
| metabolism, Proteases & Other Enzymes | |
| 95.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction