missing translation for 'onlineSavingsMsg'
Learn More

Aminoacylase/ACY1 Antibody (128), DyLight 550, Novus Biologicals™

Product Code. 18364500 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364500 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364500 Supplier Novus Biologicals Supplier No. NBP289559R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Aminoacylase/ACY1 Monoclonal specifically detects Aminoacylase/ACY1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aminoacylase/ACY1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 128
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Aminoacylase/ACY1 (NP_000657.1; Met1-Ser408).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 95
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.