missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoacylase/ACY1 Antibody (006), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Aminoacylase/ACY1 Monoclonal specifically detects Aminoacylase/ACY1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Aminoacylase/ACY1 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 6 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Aminoacylase/ACY1 (NP_000657.1; Met1-Ser408). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?