missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetonitrile sulfate, 97%

Product Code. 11369759
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369759 500 g 500g
11349759 25 g 25g
11359759 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369759 Supplier Thermo Scientific Alfa Aesar Supplier No. B22155.36
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5466-22-8 | C4H8N8·H2SO4

Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-22-8
Molecular Formula C4H8N8·H2SO4
MDL Number MFCD00012851
Synonym Glycinonitrile sulfate

Specifications

Melting Point ∼160°C (decomposition)
Odor Odorless
Linear Formula (H2NCH2CN)2·H2SO4
Quantity 500 g
UN Number UN3439
Beilstein 3916094
Sensitivity Hygroscopic
Merck Index 14,412
Solubility Information Soluble in water.
Formula Weight 210.21
Percent Purity 97%
Chemical Name or Material Aminoacetonitrile sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.