missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetonitrile bisulfate, 98%

Product Code. 10143490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143490 5 g Glass bottle 5g
10542361 100 g Glass bottle 100g
10647294 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10143490 Supplier Thermo Scientific Acros Supplier No. 153720050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 151-63-3 | C2H4N2·H2SO4 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-63-3
Molecular Formula C2H4N2·H2SO4
Molecular Weight (g/mol) 154.14
MDL Number MFCD00012851
InChI Key GTGIXCPOLMWQTC-UHFFFAOYSA-N
Synonym aminoacetonitrile bisulfate, 2-aminoacetonitrile sulfate, aminoacetonitrile hydrogen sulfate, aminoacetonitrile sulfate 1:1, aminoacetonitrile hydrogensulfate, 2-aminoacetonitrile; sulfuric acid, aminoacetonitrile bisulphate, unii-7xa82hgq1c, aminoacetonitrile hydrosulfate, aminomethylcyanide hydrogen sulfate
PubChem CID 196971
IUPAC Name 2-aminoacetonitrile;sulfuric acid
SMILES C(C#N)N.OS(=O)(=O)O

Specifications

Melting Point 123.0°C to 125.0°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NCH2CN·H2SO4
Quantity 5 g
Beilstein 04,344
Merck Index 15,406
Formula Weight 154.14
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Aminoacetonitrile bisulfate
Show More Show Less
compliance-icons
Product Identifier
  • Aminoacetonitrile bisulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.