missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetaldehyde dimethyl acetal, 99%

Product Code. 11462887
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462887 500 g 500g
11442887 25 g 25g
11452887 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462887 Supplier Thermo Scientific Alfa Aesar Supplier No. A15498.36
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22483-09-6 | C4H11NO2 | 105.137 g/mol

Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amino acetaldehyde dimethyl was used in preparation of chitosan- dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

Solubility
Miscible in water. Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22483-09-6
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00008135
InChI Key QKWWDTYDYOFRJL-UHFFFAOYSA-N
Synonym aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr
PubChem CID 89728
IUPAC Name 2,2-dimethoxyethanamine
SMILES COC(CN)OC

Specifications

Density 0.973
Boiling Point 135°C to 138°C
Flash Point 41°C (106°F)
Refractive Index 1.417
Quantity 500 g
UN Number UN2733
Beilstein 741868
Solubility Information Miscible in water. Soluble in chloroform,methanol.
Formula Weight 105.14
Percent Purity 99%
Chemical Name or Material Aminoacetaldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.