missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetaldehyde Diethyl Acetal, TRC

Product Code. 30328661
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328661 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328661 Supplier Toronto Research Chemicals Supplier No. TRCA57900025ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminoacetaldehyde Diethyl Acetal
CAS 645-36-3
Molecular Formula C6H15NO2
Quantity 25 mL
Synonym Amino-acetaldehyde Diethyl Acetal, 1,1-Diethoxy-2-aminoethane, 1-Amino-2,2-diethoxyethane, 2,2-Bis(ethyloxy)ethanamine, 2,2-Diethoxyethanamine, 2,2-Diethoxyethylamine, 2-Aminoacetaldehyde Diethyl Acetal, Aminoacetaldehyde Diethyl Acetal, Glycinaldehyde Diethyl Acetal, NSC 19501, alpha-Aminoacetaldehyde Diethyl Acetal, beta,beta-Diethoxyethylamine, 2,2-Diethoxy-ethanamine
InChI Formula InChI=1S/C6H15NO2/c1-3-8-6(5-7)9-4-2/h6H,3-5,7H2,1-2H3
SMILES CCOC(CN)OCC
IUPAC Name 2,2-diethoxyethanamine
Molecular Weight (g/mol) 133.19
Formula Weight 133.1103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.