missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetaldehyde Diethyl Acetal, 99%

Product Code. 10326371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10326371 5 mL Glass bottle 5mL
10756052 50 mL Glass bottle 50mL
10237253 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10326371 Supplier Thermo Scientific Acros Supplier No. 103070050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 645-36-3 | C6H15NO2 | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-36-3
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008136
InChI Key HJKLEAOXCZIMPI-UHFFFAOYSA-N
Synonym 2,2-diethoxyethylamine, aminoacetaldehyde diethyl acetal, ethanamine, 2,2-diethoxy, 2,2-diethoxyethan-1-amine, 2-aminoacetaldehyde diethyl acetal, glycinaldehyde diethyl acetal, aminoacetal, diethoxyethylamine, acetaldehyde, amino-, diethyl acetal, aminoacetaldehydediethylacetal
PubChem CID 69524
IUPAC Name 2,2-diethoxyethanamine
SMILES CCOC(CN)OCC

Specifications

Density 0.9160g/mL
Boiling Point 162.0°C to 163.0°C
Flash Point 45°C
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NCH2CH(OC2H5)2
Refractive Index 1.4160 to 1.4180
Quantity 5 mL
Beilstein 04,308
Specific Gravity 0.916
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ether and chloroform
Formula Weight 133.19
Percent Purity 99%
Chemical Name or Material Aminoacetaldehyde diethyl acetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aminoacetaldehyde diethyl acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.