missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetaldehyde diethyl acetal, 98%

Product Code. 11458633
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458633 100 g 100g
11448633 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458633 Supplier Thermo Scientific Alfa Aesar Supplier No. A10427.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 645-36-3 | C6H15NO2 | 133.19 g/mol

Aminoacetaldehyde diethyl acetal, is used as an agrochemical intermediate, pharmaceutical, syntheses material intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoacetaldehyde diethyl acetal, is used as an agrochemical intermediate, pharmaceutical, syntheses material intermediate.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store in a cool, dry place. Store in a tightly closed container.Take measures to prevent the build up of electrostatic charge. Stable under recommended storage conditions. Incompatabile to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, Carbon dioxide (CO2).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-36-3
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008136
InChI Key HJKLEAOXCZIMPI-UHFFFAOYSA-N
Synonym 2,2-diethoxyethylamine, aminoacetaldehyde diethyl acetal, ethanamine, 2,2-diethoxy, 2,2-diethoxyethan-1-amine, 2-aminoacetaldehyde diethyl acetal, glycinaldehyde diethyl acetal, aminoacetal, diethoxyethylamine, acetaldehyde, amino-, diethyl acetal, aminoacetaldehydediethylacetal
PubChem CID 69524
IUPAC Name 2,2-diethoxyethanamine
SMILES CCOC(CN)OCC

Specifications

Density 0.916
Boiling Point 162°C to 163°C
Flash Point 45°C (113°F)
Linear Formula H2NCH2CH(OCH2CH3)2
Refractive Index 1.417
Quantity 100 g
UN Number UN2733
Beilstein 506067
Solubility Information Soluble in water.
Formula Weight 133.19
Percent Purity 98%
Chemical Name or Material Aminoacetaldehyde diethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.