missing translation for 'onlineSavingsMsg'
Learn More

Amino-PEG3-C2-Azido, TRC

Product Code. 30326276
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
This item is not returnable. View return policy

Product Code. 30326276

Brand: Toronto Research Chemicals TRCA58500010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Amino-11-azido-3,6,9-trioxaundecane
CAS 134179-38-7
Molecular Formula C8 H18 N4 O3
Quantity 10 g
Synonym 1-Amino-11-azido-3,6,9-trioxaundecane
InChI Formula InChI=1S/C8H18N4O3/c9-1-3-13-5-7-15-8-6-14-4-2-11-12-10/h1-9H2
SMILES NCCOCCOCCOCCN=[N+]=[N-]
IUPAC Name 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethanamine
Molecular Weight (g/mol) 218.25
Formula Weight 218.1379
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.