missing translation for 'onlineSavingsMsg'
Learn More

Amino(diphenyl)acetic Acid (2,2-Diphenylglycine), Mikromol™

Product Code. 30460482
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460482 4 x 25 mg 100mg
17967864 25 mg 25mg
17908054 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30460482 Supplier Mikromol Supplier No. MM0630.0400254AMP
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amino(diphenyl)acetic Acid (2,2-Diphenylglycine), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amino(diphenyl)acetic Acid (2,2-Diphenylglycine)
CAS 3060-50-2
Analyte or Component Names Amino(diphenyl)acetic Acid (2,2-Diphenylglycine)
Shipping Condition Room Temperature
Molecular Formula C14 H13 N O2
Certifications/Compliance ISO 17025
Synonym Phenytoin Sodium Imp. C (EP), Phenytoin Imp. C (EP), Phenytoin USP Related Compound A, Phenytoin USP RC A, Amino(diphenyl)acetic Acid, 2,2-Diphenylglycine, Phenytoin Related Compound A, Phenytoin Impurity C, Phenytoin Sodium Impurity C, Phenytoin Impurity C
InChI Formula InChI=1S/C14H13NO2/c15-14(13(16)17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,15H2,(H,16,17)
Solution Type Neat
SMILES NC(C(=O)O)(c1ccccc1)c2ccccc2
IUPAC Name 2-amino-2,2-diphenylacetic acid
Molecular Weight (g/mol) 227.26
Quantity 4 x 25 mg
Formula Weight 227.0946 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.