missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amino(diphenyl)acetic Acid (2,2-Diphenylglycine), Mikromol™
Description
Amino(diphenyl)acetic Acid (2,2-Diphenylglycine)
Specifications
Specifications
| Chemical Name or Material | Amino(diphenyl)acetic Acid (2,2-Diphenylglycine) |
| CAS | 3060-50-2 |
| Analyte or Component Names | Amino(diphenyl)acetic Acid (2,2-Diphenylglycine) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H13 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Phenytoin Sodium Imp. C (EP), Phenytoin Imp. C (EP), Phenytoin USP Related Compound A, Phenytoin USP RC A, Amino(diphenyl)acetic Acid, 2,2-Diphenylglycine, Phenytoin Related Compound A, Phenytoin Impurity C, Phenytoin Sodium Impurity C, Phenytoin Impurity C |
| InChI Formula | InChI=1S/C14H13NO2/c15-14(13(16)17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,15H2,(H,16,17) |
| SMILES | NC(C(=O)O)(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2-amino-2,2-diphenylacetic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?