missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Amino Acid Diet, Synthetic, Pellet

Product Code. 11487341 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
20 kg
50 kg
Unit Size:
20kg
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487341 50 kg 50kg
11477341 20 kg 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487341 Supplier MP Biomedicals Supplier No. 0296036250
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diet rich in Amino Acids. Can be added into agar gel to increase palatability.

  • Diet rich in amino acids
  • Can be added into agar gel to increase palatability
  • Can be used for research

Specifications

Autoclavable Autoclavable
Product Type Animal Diet
Disposable Single-use
Format Pellet
Quantity 50 kg
Sterility Non-sterile
Storage Requirements Refrigerated
Length (English) 1 ft.
Length (Metric) 0.3048 m

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.