missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiloride-sensitive cation channel 3 Goat anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
£426.00
Specifications
| Antigen | Amiloride-sensitive cation channel 3 |
|---|---|
| Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Amiloride-sensitive cation channel 3 Polyclonal specifically detects Amiloride-sensitive cation channel 3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Amiloride-sensitive cation channel 3 | |
| Western Blot | |
| Unconjugated | |
| Goat | |
| Neuroscience | |
| Whole antisera | |
| 9311 | |
| IgG | |
| Unpurified | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| Mouse | |
| Acid-sensing ion channel 3, amiloride-sensitive cation channel 3, amiloride-sensitive cation channel 3, testis, ASIC3hTNaC1, DRASIC, hASIC3, modulatory subunit of ASIC2a, Neuronal amiloride-sensitive cation channel 3, proton-gated cation channel subunit, SLNAC1, Testis sodium channel 1, TNAC1 | |
| A synthetic peptide from the extracellular domain of rat amiloride sensitive cation channel 3 (ACCN3, ASIC3, SLNAC1, TNAC1) conjugated to an immunogenic carrier protein was used as the antigen. | |
| Primary | |
| Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title