missing translation for 'onlineSavingsMsg'
Learn More

Amiloride hydrochloride, MedChemExpress

Product Code. 16462086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462086 500 mg 500mg
16448216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462086 Supplier Medchem Express Supplier No. HYB0285A
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amiloride hydrochloride (MK-870 hydrochloride) is an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA). Amiloride hydrochloride is a blocker of polycystin-2 (PC2; TRPP2) channel.

Amiloride hydrochloride (MK-870 hydrochloride) is an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA). Amiloride hydrochloride is a blocker of polycystin-2 (PC2

Specifications

Chemical Name or Material Amiloride hydrochloride
CAS 2016-88-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C6H9Cl2N7O
Quantity 500 mg
Synonym MK-870 hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (375.81 mM) ∣H2O : 7.14 mg/mL (26.83 mM; Need ultrasonic)
SMILES O=C(C1=NC(Cl)=C(N)N=C1N)NC(N)=N.Cl
Molecular Weight (g/mol) 266.09
Formula Weight 266.09
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.