missing translation for 'onlineSavingsMsg'
Learn More

Amiloride hydrochloride dihydrate, MedChemExpress

Product Code. 16458216
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458216 Supplier Medchem Express Supplier No. HYB0285B100mg
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amiloride hydrochloride dihydrate (MK-870 hydrochloride dihydrate) is an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA). Amiloride hydrochloride dihydrate is a blocker of polycystin-2 (PC2; TRPP2) channel.

Amiloride hydrochloride dihydrate (MK-870 hydrochloride dihydrate) is an inhibitor of both epithelial sodium channel (ENaC) and urokinase-type plasminogen activator receptor (uTPA). Amiloride hydrochloride dihydrate is a blocker of polycystin-2 (PC2

Specifications

Chemical Name or Material Amiloride hydrochloride dihydrate
CAS 17440-83-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C6H13Cl2N7O3
Quantity 100 mg
Synonym MK-870 hydrochloride dihydrate
Solubility Information DMSO : 83.33 mg/mL (275.82 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2 mg/mL (6.62 mM; Need ultrasonic)
SMILES O=C(C1=NC(Cl)=C(N)N=C1N)NC(N)=N.[H]Cl.O.O
Molecular Weight (g/mol) 302.12
Formula Weight 302.12
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.