missing translation for 'onlineSavingsMsg'
Learn More

Amiloride EP Impurity A, Mikromol™

Product Code. 30051932
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051932 25 mg 25mg
30049861 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051932 Supplier Mikromol Supplier No. MM0560.010025
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amiloride EP Impurity A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate
CAS 1458-01-1
Analyte or Component Names Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C6 H7 Cl N4 O2
Certifications/Compliance ISO 17025
Synonym Amiloride Hydrochloride Imp. A (EP), Amiloride Imp. A (EP), Amiloride USP Related Compound A, Amiloride USP RC A, Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate, Amiloride Related Compound A (USP), 3,5-Diamino-6-chloropyrazine-2-carboxylic Acid Methyl Ester, Amiloride impurity A (EP), Amiloride Related Compound A, Amiloride Hydrochloride Dihydrate Impurity A, Amiloride Impurity A
InChI Formula InChI=1S/C6H7ClN4O2/c1-13-6(12)2-4(8)11-5(9)3(7)10-2/h1H3,(H4,8,9,11)
SMILES COC(=O)c1nc(Cl)c(N)nc1N
IUPAC Name methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
Molecular Weight (g/mol) 202.6
Quantity 25 mg
Formula Weight 202.0258 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.