missing translation for 'onlineSavingsMsg'
Learn More

Amikacin, MP Biomedicals™

Product Code. 17610877 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17610877 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17610877 Supplier MP Biomedicals Supplier No. 02150342.2
  • ONLINE EXCLUSIVE PRICE
    £64.30
    £60.27 / 250mg
    Save £4.03
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to treat different types of bacterial infections. Amikacin, MP Biomedicals is an aminoglycoside antibiotic most often used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Ente

Amikacin works by binding to the bacterial 30S ribosomal subunit, inducing misreading of mRNA and leaving the bacterium unable to synthesize proteins vital to its growth. It interferes with the translocation of tRNA from the A-site to the P-site. Amikacin must be given by the intravenous or intramuscular route; it is not absorbed orally.

Specifications

Physical Form Powder
CAS 37517-28-5
pH 9.5 to 11.5 (1% aq. soln.)
Potency ca. 900μg/mg
Color White to Light Gray
Quantity 250 mg
Optical Rotation 100.5°
Molecular Formula C22H43N5O13
Boiling Point 981.8°C (760mmHg)
Synonym 1-N-[L(-)-4-Amino-2-hydroxy-butyryl] kanamycin A, O-3-Amino-3-deoxy-a -D-glucopyranosyl-(1 ---> 6)-O-6-amino-6-deoxy-a-D-glucopyranosyl-(1 ---> 4)]- N1 -(4-amino-2-hydroxy-1-oxobutyl)-2-deoxy-D-streptamine, Lukadin
Solubility Information Sparingly soluble in water.
InChI Key LKCWBDHBTVXHDL-RMDFUYIESA-N
SMILES C1C(C(C(C(C1NC(=O)C(CCN)O)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(C(C(C(O3)CN)O)O)O)N
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
Molecular Weight (g/mol) 585.608
PubChem CID 37768
ChEBI CHEBI:2637
Formula Weight 585.6
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.