missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amikacin disulfate, MedChemExpress
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Amikacin disulfate is bactericidal, acting directly on the 30S and 50S bacerial ribosomal subunits to inhibit protein synthesis. Amikacin disulfate is very active against most Gram-negative bacteria including gentamicin- and tobramycin-resistant strains. Amikacin disulfate also inhibits the infections caused by susceptible Nocardia and nontuberculous mycobacteria.
Specifications
Specifications
| Chemical Name or Material | Amikacin disulfate |
| CAS | 39831-55-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H47N5O21S2 |
| Quantity | 1 g |
| Synonym | BAY 41-6551 disulfate |
| Solubility Information | H2O : 100 mg/mL (127.92 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| SMILES | OS(O)(=O)=O.OS(O)(=O)=O.O[C@@H]1[C@H]([C@@H](C[C@H](N)[C@H]1O[C@@]([C@@H]([C@@H](O)[C@@H]2O)O)([H])O[C@@H]2CN)NC([C@@H](O)CCN)=O)O[C@@]([C@@H]([C@@H](N)[C@@H]3O)O)([H])O[C@@H]3CO |
| Molecular Weight (g/mol) | 781.76 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction