missing translation for 'onlineSavingsMsg'
Learn More

Melford Amikacin Disulfate

Product Code. 30018010
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018010 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018010 Supplier Melford Supplier No. A20540100.0
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amikacin disulfate is an antibiotic that belongs to the aminoglycoside class.

Amikacin disulfate is an antibiotic that belongs to the aminoglycoside class. It is commonly used in microbiology and cell culture applications. This antibiotic is a broad-spectrum aminoglycoside antibiotic with potent antibacterial activity against a wide range of Gram-negative and some Gram-positive bacteria. The disulfate form of amikacin is a salt derivative, and it is often utilized in laboratory research.

Used in microbiology experiments to inhibit the growth of bacteria in cell cultures or to select for cells containing antibiotic resistance genes. It is particularly effective against certain strains of bacteria that are resistant to other antibiotics.

Researchers may use amikacin disulfate in studies involving bacterial infections, antibiotic susceptibility testing, and other experiments where selective inhibition of bacterial growth is required.

Specifications

Chemical Name or Material Amikacin Disulfate
CAS 39831-55-5
Quantity 100 g
Molecular Formula C22H43N5O13-2H2SO4
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.