Learn More
AMIGO3 Antibody (CL11023) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07991AF750
Product Code. 30302074
  • £558.00 / 0.10mL
Description
AMIGO3 Monoclonal antibody specifically detects AMIGO3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AMIGO3
Monoclonal
Alexa Fluor 750
adhesion molecule with Ig-like domain 3, ALI3, Alivin-3, AMIGO-3, amphoterin-induced gene and ORF 3, amphoterin-induced protein 3, KIAA1851, MGC120552
This antibody was developed using a synthetic peptide derived from Q86WK7, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Western Blot
CL11023
50mM Sodium Borate
Mouse
Protein A purified
RUO
386724
Store at 4⪚C in the dark.
IgG1