missing translation for 'onlineSavingsMsg'
Learn More

Amifostine thiol dihydrochloride, MedChemExpress

Product Code. 16473878
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16473878

Brand: Medchem Express HY103640

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amifostine thiol (WR-1065) dihydrochloride can protect normal tissues from the toxic effects of certain cancer drugs and activate p53 through a JNK-dependent signaling pathway.

Amifostine thiol (WR-1065) dihydrochloride can protect normal tissues from the toxic effects of certain cancer drugs and activate p53 through a JNK-dependent signaling pathway.

Specifications

Chemical Name or Material Amifostine thiol dihydrochloride
Physical Form Solid
CAS 14653-77-1
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C5H16Cl2N2S
For Use With (Application) Cancer-programmed cell death
Synonym WR-1065 dihydrochloride
Solubility Information H2O : ≥ 100 mg/mL (482.72 mM) ∣DMSO : 25 mg/mL (120.68 mM; Need ultrasonic)
SMILES NCCCNCCS.Cl.Cl
Molecular Weight (g/mol) 207.16
Formula Weight 207.16
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.