missing translation for 'onlineSavingsMsg'
Learn More

Amidotrizoic Acid, TRC

Product Code. 30137452
Change view
Click to view available options
Quantity:
250 mg
5 g
100 g
Unit Size:
100g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137452 100 g 100g
17993715 250 mg 250mg
17924395 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137452 Supplier Toronto Research Chemicals Supplier No. TRCA576800100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-96-4
Molecular Formula C11 H9 I3 N2 O4
Molecular Weight (g/mol) 613.91
Synonym Benzoic acid, 3,5-diacetamido-2,4,6-triiodo- (6CI,8CI), 3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid, 2,4,6-Triiodo-3,5-diacetamidobenzoic acid, 3,5-Diacetamido-2,4,6-triiodobenzoic acid, Amidotrizoic acid, Diatrizoate, Diatrizoic acid, NSC 262168, Odiston, Urografin acid, Urogranoic acid, Urotrast
IUPAC Name 3,5-diacetamido-2,4,6-triiodobenzoic acid
SMILES CC(=O)Nc1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I

Specifications

Analyte or Component Names Diatrizoic acid
Shipping Condition Room Temperature
InChI Formula InChI=1S/C11H9I3N2O4/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18/h1-2H3,(H,15,17)(H,16,18)(H,19,20)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 g
Formula Weight 613.7696
Chemical Name or Material Diatrizoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.