missing translation for 'onlineSavingsMsg'
Learn More

Amidotrizoic Acid Dihydrate, TRC

Product Code. 30003492
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003492 5 g 5g
30004801 25 g 25g
30006053 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003492 Supplier Toronto Research Chemicals Supplier No. TRCA5768055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50978-11-5
Molecular Formula C11 H9 I3 N2 O4 . 2 H2 O
Molecular Weight (g/mol) 649.94
Synonym Amidotrizoic acid dihydrate, 3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid dihydrate
IUPAC Name 3,5-diacetamido-2,4,6-triiodobenzoic acid;dihydrate
SMILES O.O.CC(=O)Nc1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I

Specifications

Analyte or Component Names Amidotrizoic acid dihydrate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C11H9I3N2O4.2H2O/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18;;/h1-2H3,(H,15,17)(H,16,18)(H,19,20);2*1H2
Solution Type Neat
Industry Type Forensics
Quantity 5 g
Formula Weight 649.7908
Chemical Name or Material Amidotrizoic acid dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.