missing translation for 'onlineSavingsMsg'
Learn More

Amidosulfuron, TRC

Product Code. 30132996
Change view
Click to view available options
Quantity:
100 mg
500 mg
10 g
Unit Size:
100mg
10g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132996 500 mg 500mg
30133648 10 g 10g
17960405 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132996 Supplier Toronto Research Chemicals Supplier No. TRCA576795500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120923-37-7
Molecular Formula C9 H15 N5 O7 S2
Molecular Weight (g/mol) 369.37
Synonym 2,4-Dithia-3,5-diazahexan-6-amide, N-(4,6-dimethoxy-2-pyrimidinyl)-3-methyl-, 2,2,4,4-tetraoxide (9CI), Adret, Amidosulfuron, Cratil, Gratil, Grodyl, HOE 075032, Hoestar
IUPAC Name 1-(4,6-dimethoxypyrimidin-2-yl)-3-[methyl(methylsulfonyl)sulfamoyl]urea
SMILES COc1cc(OC)nc(NC(=O)NS(=O)(=O)N(C)S(=O)(=O)C)n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H15N5O7S2/c1-14(22(4,16)17)23(18,19)13-9(15)12-8-10-6(20-2)5-7(11-8)21-3/h5H,1-4H3,(H2,10,11,12,13,15)
Formula Weight 369.0413
Chemical Name or Material Amidosulfuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.