missing translation for 'onlineSavingsMsg'
Learn More

Amidosulfonic Acid, TRC

Product Code. 30006178
Change view
Click to view available options
Quantity:
1 g
10 g
250 g
Unit Size:
10g
1g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006178 1 g 1g
30005179 10 g 10g
30011401 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006178 Supplier Toronto Research Chemicals Supplier No. TRCA5767851G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3 N O3 S
Molecular Weight (g/mol) 97.09
Synonym Sulfamic acid, Alprojet W, Amidosulfonic acid, Amidosulfuric acid, Aminesulfonic acid, Aminosulfonic acid, Aminosulfuric acid, Jumbo, NSC 1871, Scale Cleen, Steradent Active Plus, Sulfamidic acid, Sulfaminic acid, Sulphamic acid
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

Analyte or Component Names Amidosulfonic Acid
Shipping Condition Room Temperature
InChI Formula InChI=1S/H3NO3S/c1-5(2,3)4/h(H3,1,2,3,4)
Solution Type Neat
Industry Type Forensics
Quantity 1 g
Formula Weight 96.9834
Percent Purity >95%
Chemical Name or Material Amidosulfonic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.