missing translation for 'onlineSavingsMsg'
Learn More

Amidosulfonic acid, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11435240
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
5329-14-6
H3NO3S
97.088
MFCD00011603
IIACRCGMVDHOTQ-UHFFFAOYSA-N
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
5987
CHEBI:9330
sulfamic acid
NS(=O)(=O)O
This item is not returnable. View return policy
5329-14-6
H3NO3S
97.088
MFCD00011603
IIACRCGMVDHOTQ-UHFFFAOYSA-N
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
5987
CHEBI:9330
sulfamic acid
NS(=O)(=O)O

Amidosulfonic acid is mainly a precursor to sweet-tasting compounds. Reaction with cyclohexylamine followed by addition of NaOH gives C6H11NHSO3Na, sodium cyclamate. Sulfamic acid is used as an acidic cleaning agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
pH 1.2
Melting Point ∼205°C (decomposition)
Density 2.12 g/mL
Odor Odorless
Quantity 500 g
Physical Form Crystalline
UN Number UN2967
Merck Index 148921
Solubility Information Soluble in water.
Formula Weight 97.09
Percent Purity 99.99%
Assay Metals basis
Chemical Name or Material Amidosulfonic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'