missing translation for 'onlineSavingsMsg'
Learn More

Amidinothiourea, 99+%

Product Code. 10184910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10184910 10 g Glass bottle 10g
10053270 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10184910 Supplier Thermo Scientific Acros Supplier No. 259190100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2114-02-5 | C2H6N4S | 118.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2114-02-5
Molecular Formula C2H6N4S
Molecular Weight (g/mol) 118.16
MDL Number MFCD00014472
InChI Key OKGXJRGLYVRVNE-UHFFFAOYSA-N
Synonym amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
PubChem CID 2724563
IUPAC Name diaminomethylidenethiourea
SMILES C(=NC(=S)N)(N)N

Specifications

Melting Point 160°C to 164°C
Color White
Loss on Drying 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Iodimetry)
Packaging Glass bottle
Linear Formula NH2C(=NH)NHC(=S)NH2
Quantity 10 g
Solubility Information Solubility in water: 7 w/w % in water (20°C)
Formula Weight 118.16
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Amidinothiourea, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Amidinothiourea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.