missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMID Monoclonal Antibody (3A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522795
This item is not returnable.
View return policy
Description
Peptide Sequence: VRQEVKEILL RKGVQLLLSE RVSNLEELPL NEYREYIKVQ TDKGTEVATN LVILCTGIKI NSSAYRKAFE SRLASSGALR VNEHLQVEGH SNVYAIGDCA.
The p53 tumor suppressor protein induces cell cycle arrest or apoptosis in response to cellular stresses. PRG3 (p53-responsive gene 3), is induced specifically under p53-dependent apoptotic conditions in human colon cancer cells. PRG3 has significant homology to bacterial oxidoreductases and the apoptosis-inducing factor, AIF. Unlike AIF, PRG3 localizes in the cytoplasm and its ectopic expression induces apoptosis. The PRG3 gene is thus a novel p53 target gene in a p53-dependent apoptosis pathway.
Specifications
| AMID | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BRQ8 | |
| AIFM2 | |
| AIFM2 (NP_116186, 188 a.a. ~ 287 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunoprecipitation | |
| 3A11 | |
| Unconjugated | |
| AIFM2 | |
| 5430437E11Rik; AIFM2; Amid; apoptosis inducing factor, mitochondria associated 2; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; apoptosis-inducing factor 2; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated 2; apoptosis-inducing factor, mitochondrion-associated, 2; apoptosis-inducing factor-like mitchondrion-associated inducer of death; apoptosis-inducing factor-like mitochondrion-associated inducer of death; D730001I10Rik; MBP2; MBPH; MGC126662; MGC141971; p53-responsive gene 3 protein; PRG3; RP11-367H5.2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84883 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction