missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AMID Antibody (HR231), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | AMID |
|---|---|
| Clone | HR231 |
| Dilution | Western Blot : 1-5 μg/mL, ELISA, Immunohistochemistry, Immunoprecipitation |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067021
|
Novus Biologicals™
NBP3-30819-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067029
|
Novus Biologicals™
NBP3-30819-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMID Monoclonal antibody specifically detects AMID in Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western BlotSpecifications
| AMID | |
| Western Blot : 1-5 μg/mL, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Monkey, Mouse, Rat | |
| AMID, apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death, apoptosis-inducing factor 2, Apoptosis-inducing factor homologous mitochondrion-associated inducer of death, apoptosis-inducing factor, mitochondrion-associated, 2, Apoptosis-inducing factor-like mitochondrion-associated inducer of death, DKFZp686L1298, FLJ14497, p53-responsive gene 3 protein, PRG3apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer ofdeath, RP11-367H5.2 | |
| Hybridoma produced by the fusion of splenocytes from immunized with full length human AMID protein and mouse myeloma cells. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| HR231 | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Tumor Suppressors | |
| Phosphate buffered saline | |
| 84883 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title