missing translation for 'onlineSavingsMsg'
Learn More

AMID Antibody, FITC, Novus Biologicals™

Product Code. 18340822
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340822 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340822 Supplier Bio-Techne Supplier No. NBP318133F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AMID Polyclonal antibody specifically detects AMID in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMID
Applications ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias AMID, apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death, apoptosis-inducing factor 2, Apoptosis-inducing factor homologous mitochondrion-associated inducer of death, apoptosis-inducing factor, mitochondrion-associated, 2, Apoptosis-inducing factor-like mitochondrion-associated inducer of death, DKFZp686L1298, FLJ14497, p53-responsive gene 3 protein, PRG3apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer ofdeath, RP11-367H5.2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the center of human AMID. The immunogen is located within 130-180 amino acids of AMID.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 84883
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.