missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Ultra-4 Centrifugal Filter Units

Product Code. 10131842 Shop All Merck Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
Quantity:
24Pack
8 Pack
8Pack
96 Pack
96Pack
Diameter (Metric):
17.2 mm
17.3 mm
22.7 mm
Unit Size:
Pack of 24
Pack of 8
Pack of 96
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Quantity Diameter (Metric) unitSize
10131842 50 kDa 8Pack 22.7 mm Pack of 8
10191813 3 kDa 8Pack 22.7 mm Pack of 8
10384823 3 kDa 24Pack 17.3 mm Pack of 24
10283383 3 kDa 96Pack 17.3 mm Pack of 96
10640753 10 kDa 8 Pack 22.7 mm Pack of 8
10341782 10 kDa 24Pack 17.2 mm Pack of 24
10158911 10 kDa 96 Pack 17.3 mm Pack of 96
10677872 30 kDa 8Pack 22.7 mm Pack of 8
10687872 30 kDa 24Pack 22.7 mm Pack of 24
10254762 50 kDa 24Pack 22.7 mm Pack of 24
10540962 100 kDa 8Pack 22.7 mm Pack of 8
10638262 100 kDa 24Pack 22.7 mm Pack of 24
10698062 100 kDa 96Pack 22.7 mm Pack of 96
10210342 30 kDa 96Pack 22.7 mm Pack of 96
10264762 50 kDa 96Pack 22.7 mm Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 10131842 Supplier Merck Supplier No. UFC805008
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Two microcentrifuge tubes

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material (Membrane) High-recovery Ultracel-PL Regenerated Cellulose
MWCO 50 kDa
Sterility Non-sterile
Filtration Area 3 cm2
For Use With (Application) Protein purification, protein concentration and desalting
Housing Material Regenerated Cellulose (RC)
Includes Two microcentrifuge tubes
Quantity 8Pack
Type Centrifugal Filters
Volume (Metric) 4 mL
Length (Metric) 124 mm
Diameter (Metric) 22.7 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.