missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Ultra-0.5 Centrifugal Filter Units

Product Code. 10088753 Shop All Merck Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
Quantity:
24Pack
500Pack
8Pack
96 Pack
96Pack
Unit Size:
Pack of 24
Pack of 500
Pack of 8
Pack of 96
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Quantity unitSize
10088753 10 kDa 24Pack Pack of 24
10795785 3 kDa 8Pack Pack of 8
10212584 3 kDa 24Pack Pack of 24
10704645 3 kDa 96Pack Pack of 96
10130884 3 kDa 500Pack Pack of 500
10028713 10 kDa 8Pack Pack of 8
10282244 10 kDa 96 Pack Pack of 96
10150644 10 kDa 500Pack Pack of 500
10505274 30 kDa 8Pack Pack of 8
10012584 30 kDa 24Pack Pack of 24
10256744 30 kDa 96Pack Pack of 96
10328054 30 kDa 500Pack Pack of 500
10378434 50 kDa 8Pack Pack of 8
10194304 50 kDa 24Pack Pack of 24
10695445 50 kDa 96Pack Pack of 96
10257324 50 kDa 500Pack Pack of 500
10527164 100 kDa 8Pack Pack of 8
10114694 100 kDa 24Pack Pack of 24
10388244 100 kDa 96Pack Pack of 96
10398244 100 kDa 500Pack Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 10088753 Supplier Merck Supplier No. UFC501024
Add to basket
Edge
Add to basket

Includes

Two microcentrifuge tubes

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material (Membrane) High-recovery Ultracel-PL Regenerated Cellulose
MWCO 10 kDa
Sterility Non-sterile
Filtration Area 1 cm2
For Use With (Application) Protein purification, protein concentration and desalting
Housing Material Regenerated Cellulose (RC)
Includes Two microcentrifuge tubes
Max. Force 14,000 xG for concentration spin; 1000 xG for reverse spin
Quantity 24Pack
Type Centrifugal Filters
Volume (Metric) 0.5 mL
Length (English) 0.196 in.
Length (Metric) 4.99 cm
Diameter (Metric) 10.8 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.