missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Stirred Cell Maintenance Kits (Stir Bar Assembly, 2 Seals, Tubing kit)

Product Code. 15836432 Shop All Merck Products
Change view
Click to view available options
For Use With (Equipment):
Amicon Stirred Cell (50 mL)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15836432 Amicon Stirred Cell (50 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15836432 Supplier Merck Supplier No. UFSC050MT
  • ONLINE EXCLUSIVE PRICE
    £347.00
    £310.96 / Pack of 1
    Save £36.04
    10% Off
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.
TRUSTED_SUSTAINABILITY

Specifications

Description Amicon Stirred Cell 50 mL Maintenance kit
Includes Amicon™ Stirred Cell 50 mL Maintenance Kit (stir bar assembly, 2 Ea. seals, tubing kit)
Type Ultrafiltration Equipment
For Use With (Equipment) Amicon Stirred Cell (50 mL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.