missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Pro Purification System with Ultra-0.5 Device, 50 Kda

Product Code. p-7232202 Shop All Merck Products
Click to view available options
Quantity:
24Pack
Includes:
24 Amicon Ultra NMWCO 50 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 17271822

Brand: Merck ACS505024

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.

Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.

The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
MWCO 50 kDa
Sterility Non-sterile
Housing Material Regenerated Cellulose (RC)
Includes 24 Amicon Ultra NMWCO 50 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap
Quantity 24Pack
Type Centrifugal Filters
Volume (Metric) 10 mL
Length (Metric) 12 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.