missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AMICA Polyclonal Antibody
GREENER_CHOICE

Product Code. 15557982
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15557982 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557982 Supplier Invitrogen™ Supplier No. PA547491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

In sandwich ELISAs, less than 0.3% cross-reactivity with recombinant mouse AMICA, recombinant human (rh) JAM-A, rhJAM-B, and rhJAM-C is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

AMICA transmembrane protein of the plasma membrane of leukocytes that control their migration and activation through interaction with CXADR, a plasma membrane receptor found on adjacent epithelial and endothelial cells. The interaction between both receptors mediates the activation of gamma-delta T-cells, a subpopulation of T-cells residing in epithelia and involved in tissue homeostasis and repair. Upon epithelial CXADR-binding, JAML induces downstream cell signaling events in gamma-delta T-cells through PI3-kinase and MAP kinases. It results in proliferation and production of cytokines and growth factors by T-cells that in turn stimulate epithelial tissues repair. It also controls the transmigration of leukocytes within epithelial and endothelial tissues through adhesive interactions with epithelial and endothelial CXADR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMICA
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene JAML
Gene Accession No. Q86YT9
Gene Alias adhesion molecule AMICA; adhesion molecule interacting with CXADR antigen 1; adhesion molecule, interacts with CXADR antigen 1; AMICA; Amica1; Crea7; CREA7-1; CREA7-4; Dendritic cell-specific protein CREA7; Dendritic cell-specific protein CREA7-1; dendritic-cell specific protein Crea7; dendritic-cell specific protein CREA7-1; dendritic-cell specific protein CREA7-4; Gm638; Jaml; junction adhesion molecule like; junctional adhesion molecule-like; mCrea7; UNQ722/PRO1387
Gene Symbols JAML
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML Leu20-Leu275.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120425
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.