missing translation for 'onlineSavingsMsg'
Learn More

Amibegron hydrochloride, MedChemExpress

Product Code. 16406537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16406537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16406537 Supplier Medchem Express Supplier No. HY103207
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amibegron hydrochloride is a selective β3-adrenoceptor agonist, with an EC50 of 3.5 nM for β-adrenoceptor in rat colon; Amibegron hydrochloride has anxiolytic and antidepressant activity.

Amibegron hydrochloride is a selective β3-adrenoceptor agonist, with an EC50 of 3.5 nM for β-adrenoceptor in rat colon

Specifications

Chemical Name or Material Amibegron hydrochloride
CAS 121524-09-2
Color White
Purity Grade Notes Research
Molecular Formula C22H27Cl2NO4
Quantity 5 mg
Synonym SR 58611A
Solubility Information DMSO : 100 mg/mL (227.09 mM; Need ultrasonic)
SMILES O=C(OCC)COC1=CC=C2CC[C@H](NC[C@@H](C3=CC=CC(Cl)=C3)O)CC2=C1.[H]Cl
Molecular Weight (g/mol) 440.36
Formula Weight 440.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.