missing translation for 'onlineSavingsMsg'
Learn More

AMI-1, MedChemExpress

Product Code. 16440128
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16440128

Brand: Medchem Express HY18962

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress AMI-1 is a potent, cell-permeable and reversible inhibitor of protein arginine N-methyltransferases (PRMTs), with IC50s of 8.8 μM and 3.0 μM for human PRMT1 and yeast-Hmt1p, respectively. AMI-1 exerts PRMTs inhibitory effects by blocking peptide-substrate binding.

AMI-1 is a potent, cell-permeable and reversible inhibitor of protein arginine N-methyltransferases (PRMTs), with IC50s of 8.8 μM and 3.0 μM for human PRMT1 and yeast-Hmt1p, respectively. AMI-1 exerts PRMTs inhibitory effects by blocking peptide-substrate binding.

Specifications

Chemical Name or Material AMI-1
CAS 20324-87-2
Color Coffee
Purity Grade Notes Research
Molecular Formula C21H14N2Na2O9S2
Quantity 5 mg
Solubility Information H2O : 62.5 mg/mL (113.96 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CC2=CC(S(=O)(O[Na])=O)=CC(O)=C2C=C1)NC3=CC4=CC(S(=O)(O[Na])=O)=CC(O)=C4C=C3
Molecular Weight (g/mol) 548.45
Formula Weight 548.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.