missing translation for 'onlineSavingsMsg'
Learn More

AMI-1, MedChemExpress

Product Code. 16423626
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423626 25 mg 25mg
16477658 10 mg 10mg
16440128 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16423626 Supplier Medchem Express Supplier No. HY18962

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AMI-1 is a potent, cell-permeable and reversible inhibitor of protein arginine N-methyltransferases (PRMTs), with IC50s of 8.8 μM and 3.0 μM for human PRMT1 and yeast-Hmt1p, respectively. AMI-1 exerts PRMTs inhibitory effects by blocking peptide-substrate binding.

AMI-1 is a potent, cell-permeable and reversible inhibitor of protein arginine N-methyltransferases (PRMTs), with IC50s of 8.8 μM and 3.0 μM for human PRMT1 and yeast-Hmt1p, respectively. AMI-1 exerts PRMTs inhibitory effects by blocking peptide-substrate binding.

Specifications

Chemical Name or Material AMI-1
CAS 20324-87-2
Color Coffee
Purity Grade Notes Research
Molecular Formula C21H14N2Na2O9S2
Quantity 25 mg
Solubility Information H2O : 62.5 mg/mL (113.96 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CC2=CC(S(=O)(O[Na])=O)=CC(O)=C2C=C1)NC3=CC4=CC(S(=O)(O[Na])=O)=CC(O)=C4C=C3
Molecular Weight (g/mol) 548.45
Formula Weight 548.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.