missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMHR2 Recombinant Rabbit Monoclonal Antibody (23GB3205), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | AMHR2 |
|---|---|
| Clone | 23GB3205 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088859
|
Invitrogen™
MA552564 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088585
|
Invitrogen™
MA552565 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMHR2 Recombinant Monoclonal antibody specifically detects AMHR2 in Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| AMHR2 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| Q62893 | |
| 29530 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3205 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AMH type II receptor; AMHR; Amhr2; Anti-Muellerian hormone type II receptor; anti-Muellerian hormone type-2 receptor; anti-Mullerian hormone receptor type 11 SV1; anti-Mullerian hormone receptor type 2; anti-Mullerian hormone receptor type II; anti-Mullerian hormone receptor, type II; anti-Mullerian hormone type 2 receptor; anti-Mullerian hormone type 2 receptor delta 2; anti-Mullerian hormone type 2 receptor delta 9/10; C14; MIS type II receptor; Misiir; MISR2; MISRII; MRII; Muellerian inhibiting substance type II receptor; Mullerian inhibiting substance type II receptor | |
| A synthetic peptide of human AMHR2 (77-124AA). | |
| Primary | |
| -20°C | |
| AMHR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title