missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMH Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14461-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to AMH.
Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.Specifications
| AMH | |
| Polyclonal | |
| Unconjugated | |
| AMH | |
| AMH, anti Mullerian hormone, MIF, MIS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11705, 268 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P03971, P27106 | |
| AMH | |
| AMH Fusion Protein Ag5670 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction