missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584740
This item is not returnable.
View return policy
Description
Immunogen sequence: GEDSRLSTAR LQALLFGDDH RCFTRMTPAL LLLPRSEPAP LPAHGQLDTV PFPPPRPSAE LEESPPSADP FLETLT Highest antigen sequence indentity to the following orthologs: Rat - 67%, Mouse - 66%.
Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
Specifications
| AMH | |
| Polyclonal | |
| Unconjugated | |
| AMH | |
| AMH; Anti - Mullerian hormone (Mulerian inhibiting substance); anti-Muellerian hormone; anti-Mullerian hormone; MIF; MIS; Muellerian-inhibiting factor; muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 268 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P03971 | |
| AMH | |
| Recombinant protein corresponding to Human AMH. Recombinant protein control fragment (Product #RP-108520). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction