missing translation for 'onlineSavingsMsg'
Learn More

AMG9810, MedChemExpress

Product Code. 16406267
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406267

Brand: Medchem Express HY1017365mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AMG9810 is a selective and competitive vanilloid receptor 1 (TRPV1) antagonist with IC50 values of 24.5 and 85.6 nM for human and rat TRPV1, repectively.

AMG9810 is a selective and competitive vanilloid receptor 1 (TRPV1) antagonist with IC50 values of 24.5 and 85.6 nM for human and rat TRPV1, repectively.

Specifications

Chemical Name or Material AMG9810
CAS 545395-94-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H23NO3
Quantity 5 mg
Solubility Information DMSO : ≥ 33 mg/mL (97.80 mM)
SMILES O=C(NC1=CC=C(OCCO2)C2=C1)/C=C/C3=CC=C(C(C)(C)C)C=C3
Molecular Weight (g/mol) 337.41
Formula Weight 337.41
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.